Assay Details
| Gene Symbol: CYSLTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102240.2 | NP_001095710.1 | 1 - 2 | 308 | 129 | |
| GenBank mRNA | BC151559.1 | - | 2 - 3 | 349 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151559.1, NM_001102240.2 transcripts.
Set Membership: |
|

CYSLTR2
Genomic Map