Assay Details
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102243.2 | NP_001095713.1 | 1 - 2 | 217 | 81 | |
GenBank mRNA | BC151462.1 | - | 1 - 2 | 218 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
