Assay Details
| Gene Symbol: RNF121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102245.1 | NP_001095715.1 | 6 - 7 | 739 | 71 | |
| GenBank mRNA | BC149267.1 | - | 6 - 7 | 739 | 71 | |
More Information
Set Membership: |
|

RNF121
Genomic Map