Assay Details
Gene Symbol: FOSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102248.1 | NP_001095718.1 | 2 - 3 | 471 | 116 | |
GenBank mRNA | BC148047.1 | - | 2 - 3 | 471 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
