Assay Details
| Gene Symbol: SLF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102259.1 | NP_001095729.1 | 14 - 15 | 3160 | 64 | |
| GenBank mRNA | BC151640.1 | - | 14 - 15 | 3160 | 64 | |
More Information
Set Membership: |
|

SLF2
Genomic Map