Assay Details
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102265.2 | NP_001095735.1 | 4 - 5 | 788 | 57 | |
GenBank mRNA | BC149669.1 | - | 4 - 5 | 789 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
