Assay Details
| Gene Symbol: GAS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102280.2 | NP_001095750.1 | 10 - 11 | 1252 | 55 | |
| GenBank mRNA | BC149335.1 | - | 10 - 11 | 1253 | 55 | |
| GQ500693.1 | - | 11 - 12 | 1054 | 55 | ||
More Information
Set Membership: |
|

GAS7
Genomic Map