Assay Details
| Gene Symbol: EIF4E3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102306.1 | NP_001095776.1 | 4 - 5 | 429 | 68 | |
| GenBank mRNA | BC148114.1 | - | 4 - 5 | 429 | 68 | |
More Information
Set Membership: |
|

EIF4E3
Genomic Map