Assay Details
| Gene Symbol: APOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102380.2 | NP_001095850.1 | 2 - 3 | 109 | 78 | |
| GenBank mRNA | BC146135.1 | - | 2 - 3 | 110 | 78 | |
More Information
Set Membership: |
|

APOC2
Genomic Map