Assay Details
| Gene Symbol: ACOT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103275.2 | NP_001096745.2 | 5 - 6 | 641 | 71 | |
| GenBank mRNA | BC150117.1 | - | 5 - 6 | 574 | 71 | |
| KR149245.1 | - | 5 - 6 | 574 | 71 | ||
More Information
Set Membership: |
|

ACOT11
Genomic Map