Assay Details
| Gene Symbol: DHCR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103276.1 | NP_001096746.1 | 4 - 5 | 679 | 58 | |
| GenBank mRNA | BC150073.1 | - | 4 - 5 | 679 | 58 | |
More Information
Set Membership: |
|

DHCR24
Genomic Map