Assay Details
| Gene Symbol: GTF2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103294.1 | NP_001096764.1 | 2 - 3 | 515 | 114 | |
| GenBank mRNA | BC147980.1 | - | 2 - 3 | 515 | 114 | |
More Information
Set Membership: |
|

GTF2E1
Genomic Map