Assay Details
Gene Symbol: SUPT3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105008.1 | NP_001098478.1 | 3 - 4 | 337 | 79 | |
GenBank mRNA | BC149055.1 | - | 3 - 4 | 337 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
