Assay Details
Gene Symbol: CYB5RL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105397.1 | NP_001098867.1 | 3 - 4 | 413 | 57 | |
GenBank mRNA | BC149172.1 | - | 3 - 4 | 413 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
