Assay Details
| Gene Symbol: CYB5RL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105397.1 | NP_001098867.1 | 6 - 7 | 755 | 93 | |
| GenBank mRNA | BC149172.1 | - | 6 - 7 | 755 | 93 | |
More Information
Set Membership: |
|

CYB5RL
Genomic Map