Assay Details
| Gene Symbol: WASHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105406.1 | NP_001098876.1 | 2 - 3 | 349 | 89 | |
| GenBank mRNA | BC153261.1 | - | 2 - 3 | 349 | 89 | |
More Information
Set Membership: |
|

WASHC1
Genomic Map