Assay Details
Gene Symbol: KIAA2012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105438.2 | NP_001098908.2 | 20 - 21 | 3341 | 58 | |
GenBank mRNA | BC149723.1 | - | 20 - 21 | 3344 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
