Assay Details
| Gene Symbol: ZNRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105443.2 | NP_001098913.1 | 3 - 4 | 995 | 81 | |
| GenBank mRNA | BC142537.1 | - | 3 - 4 | 996 | 81 | |
More Information
Set Membership: |
|

ZNRF2
Genomic Map