Assay Details
| Gene Symbol: DUPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105444.1 | NP_001098914.1 | 1 - 2 | 56 | 60 | |
| GenBank mRNA | BC149835.1 | - | 1 - 2 | 56 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149835.1, NM_001105444.1 transcripts.
Set Membership: |
|

DUPD1
Genomic Map