Assay Details
| Gene Symbol: CXCL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113174.1 | NP_001106645.1 | 2 - 3 | 214 | 76 | |
| GenBank mRNA | BC119979.1 | - | 2 - 3 | 214 | 76 | |
| KP057086.1 | - | 2 - 3 | 178 | 76 | ||
More Information
Set Membership: |
|

CXCL12
Genomic Map