Assay Details
| Gene Symbol: RPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113309.2 | NP_001106780.1 | 1 | 63 | 131 | |
| GenBank mRNA | BC108179.1 | - | 1 | 65 | 131 | |
| BC148023.1 | - | 2 - 3 | 68 | 131 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108179.1, BC148023.1, NM_001113309.2 transcripts.
Set Membership: |
|

RPL38
Genomic Map