Assay Details
Gene Symbol: RPL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113309.2 | NP_001106780.1 | 1 | 127 | 121 | |
GenBank mRNA | BC108179.1 | - | 1 | 129 | 121 | |
BC148023.1 | - | 3 - 4 | 132 | 121 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC108179.1, BC148023.1, NM_001113309.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
