Assay Details
| Gene Symbol: SEM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113317.2 | NP_001106788.1 | 1 | 263 | 122 | |
| GenBank mRNA | BC103152.1 | - | 1 | 264 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SEM1| Gene Symbol: SEM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113317.2 | NP_001106788.1 | 1 | 263 | 122 | |
| GenBank mRNA | BC103152.1 | - | 1 | 264 | 122 | |
Set Membership: |
|