Assay Details
| Gene Symbol: LOC780933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113727.2 | NP_001107199.1 | 3 - 4 | 481 | 81 | |
| GenBank mRNA | BC134797.1 | - | 3 - 4 | 482 | 81 | |
| D38507.1 | - | 3 - 4 | 444 | 81 | ||
| Gene Symbol: PRSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC146041.1 | - | 3 - 4 | 491 | 81 | |
More Information
Set Membership: |
|

LOC780933
Genomic Map