Assay Details
Gene Symbol: LSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114519.2 | NP_001107991.1 | 2 - 3 | 137 | 89 | |
GenBank mRNA | BC118348.1 | - | 2 - 3 | 138 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
