Assay Details
Gene Symbol: LSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114519.2 | NP_001107991.1 | 3 - 4 | 247 | 118 | |
GenBank mRNA | BC118348.1 | - | 3 - 4 | 248 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
