Assay Details
| Gene Symbol: GCHFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114526.2 | NP_001107998.1 | 1 - 2 | 151 | 85 | |
| GenBank mRNA | BC109778.1 | - | 1 - 2 | 152 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109778.1, NM_001114526.2 transcripts.
Set Membership: |
|

GCHFR
Genomic Map