Assay Details
| Gene Symbol: LOC788142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099207.2 | NP_001092677.1 | 2 - 3 | 315 | 99 | |
| GenBank mRNA | BC142057.1 | - | 2 - 3 | 316 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC142057.1, NM_001099207.2 transcripts.
Set Membership: |
|

LOC788142
Genomic Map