Assay Details
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099210.2 | NP_001092680.1 | 1 - 2 | 46 | 105 | |
GenBank mRNA | BC142374.1 | - | 1 - 2 | 48 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC142374.1, NM_001099210.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
