Assay Details
Gene Symbol: RPS17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099210.2 | NP_001092680.1 | 2 - 3 | 198 | 72 | |
GenBank mRNA | BC142374.1 | - | 2 - 3 | 200 | 72 | |
BC148018.1 | - | 2 - 3 | 182 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
