Assay Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099213.1 | NP_001092683.1 | 2 - 3 | 346 | 68 | |
| GenBank mRNA | BC142019.1 | - | 2 - 3 | 346 | 68 | |
More Information
Set Membership: |
|

TMEM170A
Genomic Map