Assay Details
| Gene Symbol: MOB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099732.1 | NP_001093202.1 | 3 - 4 | 346 | 57 | |
| GenBank mRNA | BC142101.1 | - | 3 - 4 | 346 | 57 | |
More Information
Set Membership: |
|

MOB2
Genomic Map