Assay Details
| Gene Symbol: PTER | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101282.1 | NP_001094752.1 | 3 - 4 | 845 | 102 | |
| GenBank mRNA | BC147990.1 | - | 3 - 4 | 845 | 102 | |
More Information
Set Membership: |
|

PTER
Genomic Map