Assay Details
Gene Symbol: KRT32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101302.1 | NP_001094772.1 | 4 - 5 | 830 | 68 | |
GenBank mRNA | BC149206.1 | - | 4 - 5 | 830 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
