Assay Details
| Gene Symbol: NATD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102331.2 | NP_001095801.1 | 2 - 3 | 340 | 101 | |
| GenBank mRNA | BC147869.1 | - | 2 - 3 | 342 | 101 | |
More Information
Set Membership: |
|

NATD1
Genomic Map