Assay Details
Gene Symbol: BTNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102351.1 | NP_001095821.1 | 5 - 6 | 991 | 70 | |
GenBank mRNA | BC148938.1 | - | 5 - 6 | 991 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
