Assay Details
| Gene Symbol: KHNYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102551.1 | NP_001096021.1 | 7 - 8 | 1926 | 63 | |
| GenBank mRNA | BC151400.1 | - | 7 - 8 | 1926 | 63 | |
More Information
Set Membership: |
|

KHNYN
Genomic Map