Assay Details
| Gene Symbol: CX3CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102558.2 | NP_001096028.1 | 1 - 2 | 83 | 83 | |
| GenBank mRNA | BC148890.1 | - | 1 - 2 | 84 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC148890.1, NM_001102558.2 transcripts.
Set Membership: |
|

CX3CR1
Genomic Map