Assay Details
| Gene Symbol: HACD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103316.1 | NP_001096786.1 | 3 - 4 | 321 | 78 | |
| GenBank mRNA | BC151339.1 | - | 3 - 4 | 321 | 78 | |
More Information
Set Membership: |
|

HACD3
Genomic Map