Assay Details
| Gene Symbol: LOC781423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105012.2 | NP_001098482.1 | 1 - 2 | 136 | 81 | |
| GenBank mRNA | BC149901.1 | - | 1 - 2 | 137 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149901.1, NM_001105012.2 transcripts.
Set Membership: |
|

LOC781423
Genomic Map