Assay Details
| Gene Symbol: CEP295NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105031.2 | NP_001098501.2 | 2 - 3 | 187 | 60 | |
| GenBank mRNA | BC149293.1 | - | 2 - 3 | 185 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149293.1, NM_001105031.2 transcripts.
Set Membership: |
|

CEP295NL
Genomic Map