Assay Details
| Gene Symbol: NREP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105045.1 | NP_001098515.1 | 3 - 4 | 195 | 93 | |
| GenBank mRNA | BC133421.1 | - | 3 - 4 | 195 | 93 | |
More Information
Set Membership: |
|

NREP
Genomic Map