Assay Details
| Gene Symbol: CRTAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105457.1 | NP_001098927.1 | 5 - 6 | 697 | 61 | |
| GenBank mRNA | BC134770.1 | - | 5 - 6 | 697 | 61 | |
More Information
Set Membership: |
|

CRTAM
Genomic Map