Assay Details
| Gene Symbol: SOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105463.2 | NP_001098933.1 | 1 | 1346 | 112 | |
| GenBank mRNA | BC133458.1 | - | 1 | 1347 | 112 | |
More Information
Set Membership: |
|

SOX2
Genomic Map