Assay Details
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105481.1 | NP_001098951.1 | 2 - 3 | 191 | 67 | |
GenBank mRNA | BC123755.1 | - | 2 - 3 | 191 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
