Assay Details
Gene Symbol: IRAK1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110093.1 | NP_001103563.1 | 2 - 3 | 416 | 122 | |
GenBank mRNA | BC109685.1 | - | 2 - 3 | 416 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
