Assay Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110444.2 | NP_001103914.1 | 3 - 4 | 705 | 85 | |
| GenBank mRNA | BC126697.1 | - | 3 - 4 | 706 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC126697.1, NM_001110444.2 transcripts.
Set Membership: |
|

ZNF713
Genomic Map