Assay Details
| Gene Symbol: BCAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111263.1 | NP_001104733.1 | 2 - 3 | 194 | 71 | |
| GenBank mRNA | EF446906.1 | - | 2 - 3 | 194 | 71 | |
| EF446907.1 | - | 2 - 3 | 194 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001111263.1 transcripts.
Set Membership: |
|

BCAR4
Genomic Map