Assay Details
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113572.1 | NP_001107044.1 | 2 - 3 | 406 | 67 | |
GenBank mRNA | CO878421.1 | - | 2 - 3 | 297 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
