Assay Details
Gene Symbol: BOLA-DQB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012676.3 | NP_001012694.2 | 3 - 4 | 741 | 105 | |
GenBank mRNA | BC133449.1 | - | 3 - 4 | 742 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
